(4-Aminophenyl)[4-(3-phenyl-2-propenyl)piperazino]methanone ≥95%

Supplier: Matrix Scientific

044929-500MG 044929-1G 044929-5G
101868-274EA 375.38 USD
101868-274 101868-276 101868-278
(4-Aminophenyl)[4-(3-phenyl-2-propenyl)piperazino]methanone ≥95%
(4-Aminophenyl)[4-(3-phenyl-2-propenyl)piperazino]methanone

Hazard: Irritant

Formula: C₂₀H₂₃N₃O
Melting Pt: 138 °C
MDL Number: MFCD06659559
CAS Number: 219989-07-8

Order Now

Specification Test Results

Melting point 138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR