(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone ≥95%

Supplier: Matrix Scientific

044928-500MG 044928-1G 044928-5G
101868-268EA 375.38 USD
101868-268 101868-270 101868-272
(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone ≥95%
(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone

Hazard: Irritant

Formula: C₁₅H₁₇N₅O
MW: 283.34 g/mol
Melting Pt: 216 °C
MDL Number: MFCD06659558
CAS Number: 885949-72-4

Order Now

Specification Test Results

Melting point 216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR