(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone ≥95%

Supplier: Matrix Scientific

044927-500MG 044927-1G 044927-5G
101868-262EA 375.38 USD
101868-262 101868-264 101868-266
(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone ≥95%
(4-Aminophenyl)[4-(2-fluorophenyl)piperazino]methanone

Hazard: Irritant

Formula: C₁₇H₁₈FN₃O
MW: 299.35 g/mol
Melting Pt: 161 °C
MDL Number: MFCD06659557
CAS Number: 885949-71-3

Order Now

Specification Test Results

Melting point 161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR