(4-Aminophenyl)(4-cyclohexylpiperazino)methanone ≥95%

Supplier: Matrix Scientific

044926-500MG 044926-1G 044926-5G
101868-256EA 375.38 USD
101868-256 101868-258 101868-260
(4-Aminophenyl)(4-cyclohexylpiperazino)methanone ≥95%
(4-Aminophenyl)(4-cyclohexylpiperazino)methanone

Hazard: Irritant

Formula: C₁₇H₂₅N₃O
MW: 287.41 g/mol
Melting Pt: 134 °C
MDL Number: MFCD06659556
CAS Number: 885949-70-2

Order Now

Specification Test Results

Melting point 134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR