(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone ≥95%

Supplier: Matrix Scientific

044925-500MG 044925-1G 044925-5G
101868-250EA 375.38 USD
101868-250 101868-252 101868-254
(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone ≥95%
(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone

Hazard: Irritant

Formula: C₁₆H₁₈N₄O
MW: 282.35 g/mol
Melting Pt: 138 °C
MDL Number: MFCD06659555
CAS Number: 885949-69-9

Order Now

Specification Test Results

Melting point 138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR