2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Methylthio)oxazolo[4,5-b]pyridine

044924-500MG 044924-1G 044924-5G
101868-244EA 278.46 USD
101868-244 101868-246 101868-248
2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine ≥95%
2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine

Hazard: Irritant

Formula: C₇H₆N₂OS
MW: 166.2 g/mol
Melting Pt: 59…62 °C
MDL Number: MFCD00973738
CAS Number: 169205-95-2

Order Now

Specification Test Results

Melting point 59-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR