1,3-Oxazolo[4,5-b]pyridine-2-thiol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Thio-1,3-oxazolo[4,5-b]pyridine, Oxazolo[4,5-b]pyridine-2-thiol
044922-500MG
044922-1G
044922-5G
101868-232EA
95.38
USD
101868-232
101868-234
101868-236
1,3-Oxazolo[4,5-b]pyridine-2-thiol ≥95%
1,3-Oxazolo[4,5-b]pyridine-2-thiol
Hazard: Irritant
Formula:
C₆H₄N₂OS MW: 152.174 g/mol Melting Pt: 241…243 °C |
MDL Number:
MFCD00973914 CAS Number: 211949-57-4 |
Specification Test Results
Melting point | 241-243°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...