2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid ≥95%

Supplier: Matrix Scientific

044920-500MG 044920-1G 044920-5G
101868-226EA 181.54 USD
101868-226 101868-228 101868-230
2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid ≥95%
2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₄
MW: 218.21 g/mol
Melting Pt: 129…130 °C
MDL Number: MFCD06411618
CAS Number: 624722-10-7

Order Now

Specification Test Results

Melting point 129-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR