3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-methoxy-1H-indole ≥95%

Supplier: Matrix Scientific

044919-500MG 044919-1G 044919-5G
101868-220EA 375.38 USD
101868-220 101868-222 101868-224
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-methoxy-1H-indole ≥95%
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-methoxy-1H-indole

Hazard: Irritant

Formula: C₁₇H₁₉NO₃
MW: 285.35 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD06200976
CAS Number: 185383-63-5

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR