4-(tert-Butyl)benzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

044915-500MG 044915-1G 044915-5G
101868-198EA 181.54 USD
101868-198 101868-200 101868-202
4-(tert-Butyl)benzenecarbaldehyde oxime ≥95%
4-(tert-Butyl)benzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₁₁H₁₅NO
Melting Pt: 105…107 °C
MDL Number: MFCD01595462
CAS Number: 180261-48-7

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR