3-Chloro-1H-indole-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

044914-500MG 044914-1G 044914-5G
101868-192EA 556.92 USD
101868-192 101868-194 101868-196
3-Chloro-1H-indole-2-carbohydrazide ≥95%
3-Chloro-1H-indole-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₈ClN₃O
Melting Pt: 230…232 °C
MDL Number: MFCD04125739
CAS Number: 441801-15-6

Order Now

Specification Test Results

Melting point 230-232°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR