2-Methyl-1,3-benzoxazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazol-5-amine

044912-500MG 044912-1G 044912-5G
101868-180EA 112.31 USD
101868-180 101868-182 101868-184
2-Methyl-1,3-benzoxazol-5-amine ≥95%
2-Methyl-1,3-benzoxazol-5-amine

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.161 g/mol
Melting Pt: 50…53 °C
MDL Number: MFCD00628970
CAS Number: 72745-76-7

Order Now

Specification Test Results

Melting point 50-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR