N-(3-Methoxyphenyl)-2,2-dimethylpropanamide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2,2-Dimethylpropanoyl)-3-(methoxy)aniline

044907-500MG 044907-1G 044907-5G
101868-150EA 100 USD
101868-150 101868-152 101868-154
N-(3-Methoxyphenyl)-2,2-dimethylpropanamide ≥95%
N-(3-Methoxyphenyl)-2,2-dimethylpropanamide

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Melting Pt: 125…126 °C
MDL Number: MFCD00662896
CAS Number: 56619-93-3

Order Now

Specification Test Results

Melting point 125-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR