2-[2-(2-Amino-1,3-thiazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide ≥95%
Supplier: Matrix Scientific
044905-500MG
044905-1G
044905-5G
101868-138EA
140
USD
101868-138
101868-140
101868-142
2-[2-(2-Amino-1,3-thiazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide ≥95%
2-[2-(2-Amino-1,3-thiazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
Hazard: Irritant
Formula:
C₁₃H₁₂BrN₃O₂S Melting Pt: 266…267 °C |
MDL Number:
MFCD04125720 CAS Number: 95914-09-3 |
Specification Test Results
Melting point | 266-267°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...