6-Propyl-4-(trifluoromethyl)-2-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-6-(prop-1-yl)-4-(trifluoromethyl)pyridine

044904-500MG 044904-1G
101868-134EA 556.92 USD
101868-134 101868-136
6-Propyl-4-(trifluoromethyl)-2-pyridinol ≥95%
6-Propyl-4-(trifluoromethyl)-2-pyridinol

Hazard: Irritant

Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
Melting Pt: 75…7 °C
MDL Number: MFCD08443946
CAS Number: 937601-39-3

Order Now

Specification Test Results

Melting point 75-7°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR