1-(4-Fluorophenyl)-2-imidazolidinone ≥95%
Supplier: Matrix Scientific
044902-500MG
044902-1G
044902-5G
101868-128EA
375.38
USD
101868-128
101868-130
101868-132
1-(4-Fluorophenyl)-2-imidazolidinone ≥95%
1-(4-Fluorophenyl)-2-imidazolidinone
Hazard: Irritant
Formula:
C₉H₉FN₂O Melting Pt: 150 °C |
MDL Number:
MFCD03787796 CAS Number: 53159-75-4 |
Specification Test Results
Melting point | 150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...