N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea ≥95%

Supplier: Matrix Scientific

044901-500MG 044901-1G 044901-5G
101868-122EA 278.46 USD
101868-122 101868-124 101868-126
N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea ≥95%
N-(2-Chloroethyl)-N'-(4-fluorophenyl)urea

Hazard: Irritant

Formula: C₉H₁₀ClFN₂O
Melting Pt: 132 °C
MDL Number: MFCD03787786
CAS Number: 13908-32-2

Order Now

Specification Test Results

Melting point 132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR