4-Amino-N-(2-piperidinoethyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-N-[2-(piperidin-1-yl)ethyl]benzamide

044898-500MG 044898-1G 044898-5G
101868-116EA 556.92 USD
101868-116 101868-118 101868-120
4-Amino-N-(2-piperidinoethyl)benzenecarboxamide ≥95%
4-Amino-N-(2-piperidinoethyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MW: 247.34 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD06808529
CAS Number: 51-08-1

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR