Methyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044893-500MG 044893-1G 044893-5G
101868-092EA 215.38 USD
101868-092 101868-094 101868-096
Methyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂S
Melting Pt: 223…225 °C
MDL Number: MFCD00454389
CAS Number: 115174-39-5

Order Now

Specification Test Results

Melting point 223-225°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR