Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromo-4-methylthiazole-5-carboxylate

044891-1G 044891-5G
101868-082EA 18.46 USD
101868-082 101868-084
Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥95%
Ethyl-2-bromo-4-methylthiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₈BrNO₂S
MW: 250.12 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD03791227
CAS Number: 22900-83-0

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR