1-(2-Amino-5-morpholinophenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

044889-500MG 044889-1G 044889-5G
101868-070EA 921.54 USD
101868-070 101868-072 101868-074
1-(2-Amino-5-morpholinophenyl)-1-ethanone ≥95%
1-(2-Amino-5-morpholinophenyl)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Melting Pt: 104…105 °C
MDL Number: MFCD03787743
CAS Number: 98440-50-7

Order Now

Specification Test Results

Melting point 104-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR