4-Chlorooxindole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloroindolin-2-one

044887-1G 044887-5G
101868-060EA 56.92 USD
101868-060 101868-062
4-Chlorooxindole ≥95%
4-Chlorooxindole

Hazard: Irritant

Formula: C₈H₆ClNO
MW: 167.5900 g/mol
Melting Pt: 206…208 °C
MDL Number: MFCD03787567
CAS Number: 20870-77-3

Order Now

Specification Test Results

Melting point 206-208°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR