Dimethyl-2-(4-chloro-2-nitrophenyl)malonate ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-(4-chloro-2-nitrophenyl)malonate

044886-500MG 044886-1G 044886-5G
101868-054EA 69.23 USD
101868-054 101868-056 101868-058
Dimethyl-2-(4-chloro-2-nitrophenyl)malonate ≥95%
Dimethyl-2-(4-chloro-2-nitrophenyl)malonate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₆
MW: 287.66 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD00179451
CAS Number: 147124-32-1

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR