Dimethyl-2-(2-chloro-6-nitrophenyl)malonate ≥95%

Supplier: Matrix Scientific

044885-500MG 044885-1G 044885-5G
101868-048EA 181.54 USD
101868-048 101868-050 101868-052
Dimethyl-2-(2-chloro-6-nitrophenyl)malonate ≥95%
Dimethyl-2-(2-chloro-6-nitrophenyl)malonate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₆
MW: 287.66 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD03787565
CAS Number: 147124-36-5

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR