Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-(4-chlorophenyl)-2-cyanoacrylate
044883-500MG
044883-1G
044883-5G
101868-040EA
181.54
USD
101868-040
101868-042
101868-044
Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate ≥95%
Ethyl-3-(4-chlorophenyl)-2-cyanoacrylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₂ MW: 235.67 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD00454124 CAS Number: 2286-35-3 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...