Ethyl-1-[3-(aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044879-500MG 044879-1G 044879-5G
101868-018EA 278.46 USD
101868-018 101868-020 101868-022
Ethyl-1-[3-(aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate ≥95%
Ethyl-1-[3-(aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₅
Melting Pt: 147…150 °C
MDL Number: MFCD03425790
CAS Number: 439095-47-3

Order Now

Specification Test Results

Melting point 147-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR