5-Cyano-4-methyl-3-phenyl-2-thiophenecarboxamide ≥95%

Supplier: Matrix Scientific

044878-500MG 044878-1G 044878-5G
101868-012EA 375.38 USD
101868-012 101868-014 101868-016
5-Cyano-4-methyl-3-phenyl-2-thiophenecarboxamide ≥95%
5-Cyano-4-methyl-3-phenyl-2-thiophenecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₀N₂OS
Melting Pt: 181…183 °C
MDL Number: MFCD03425789
CAS Number: 70541-99-0

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR