1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044871-500MG 044871-1G 044871-5G
101867-976EA 181.54 USD
101867-976 101867-978 101867-980
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-indole-3-carbaldehyde ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₉ClN₂OS
Melting Pt: 140…142 °C
MDL Number: MFCD03425786
CAS Number: 439095-43-9

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR