5,6-Dichloro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Dichlorobenzimidazole, 5,6-Dichloro-1H-benzo[d]imidazole

044865-500MG 044865-1G 044865-5G
101867-950EA 78.46 USD
101867-950 101867-952 101867-954
5,6-Dichloro-1H-1,3-benzimidazole ≥95%
5,6-Dichloro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
Melting Pt: 205…206 °C
MDL Number: MFCD00211325
CAS Number: 6478-73-5

Order Now

Specification Test Results

Melting point 205-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR