2-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-((2-Chlorothiazol-5-yl)methoxy)phenyl)acetonitrile
044864-500MG
044864-1G
101867-946EA
375.38
USD
101867-946
101867-948
2-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetonitrile ≥95%
2-{4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetonitrile
Hazard: Irritant
Formula:
C₁₂H₉ClN₂OS Melting Pt: 68…69 °C |
MDL Number:
MFCD02186594 CAS Number: 672950-06-0 |
Specification Test Results
Melting point | 68-69°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...