N-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetamide ≥95%

Supplier: Matrix Scientific

044863-500MG 044863-1G 044863-5G
101867-940EA 181.54 USD
101867-940 101867-942 101867-944
N-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetamide ≥95%
N-{3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetamide

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O₂S
MW: 282.75 g/mol
Melting Pt: 93 °C
MDL Number: MFCD02186592
CAS Number: 453557-52-3

Order Now

Specification Test Results

Melting point 93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR