1-Methyl-4-(5-nitropyridin-2-yl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Methyl-1-piperazinyl)-5-nitropyridine

044856-500MG 044856-1G 044856-5G
101867-914EA 278.46 USD
101867-914 101867-916 101867-918
1-Methyl-4-(5-nitropyridin-2-yl)piperazine ≥95%
1-Methyl-4-(5-nitropyridin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₄N₄O₂
MW: 222.240 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD03001270
CAS Number: 55403-34-4

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR