3-(2-Methoxyphenyl)-2-phenylacrylic acid ≥95%

Supplier: Matrix Scientific

044855-500MG 044855-1G 044855-5G
101867-908EA 278.46 USD
101867-908 101867-910 101867-912
3-(2-Methoxyphenyl)-2-phenylacrylic acid ≥95%
3-(2-Methoxyphenyl)-2-phenylacrylic acid

Hazard: Irritant

Formula: C₁₆H₁₄O₃
Melting Pt: 187…189 °C
MDL Number: MFCD01673866
CAS Number: 20890-72-6

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR