2'-(4-Fluorobenzyloxy)acetophenone ≥95%

Supplier: Matrix Scientific

044852-500MG 044852-1G 044852-5G
101867-890EA 181.54 USD
101867-890 101867-892 101867-894
2'-(4-Fluorobenzyloxy)acetophenone ≥95%
2'-(4-Fluorobenzyloxy)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₃FO₂
Melting Pt: 56…58 °C
MDL Number: MFCD03001269
CAS Number: 400878-24-2

Order Now

Specification Test Results

Melting point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR