2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044850-500MG 044850-1G 044850-5G
101867-880EA 556.92 USD
101867-880 101867-882 101867-884
2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde ≥95%
2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₀N₄O
Melting Pt: 230 °C
MDL Number: MFCD02187166
CAS Number: 453557-49-8

Order Now

Specification Test Results

Melting point 230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR