1-(4-Chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylic acid, WAY-271999

044847-500MG 044847-1G 044847-5G
101867-862EA 161.54 USD
101867-862 101867-864 101867-866
1-(4-Chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylic acid ≥95%
1-(4-Chlorophenyl)-4-oxo-1,4-dihydro-3-pyridazinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₃
MW: 250.64 g/mol
Melting Pt: 230…234 °C
MDL Number: MFCD01871546
CAS Number: 147920-34-1

Order Now

Specification Test Results

Melting point 230-234°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR