2-(4-Chlorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

044841-500MG 044841-1G 044841-5G
101867-834EA 375.38 USD
101867-834 101867-836 101867-838
2-(4-Chlorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
2-(4-Chlorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

Hazard: Irritant

Formula: C₁₀H₅ClN₄O₂
MW: 248.63 g/mol
Melting Pt: 220…224 °C
MDL Number: MFCD01316193
CAS Number: 20932-04-1

Order Now

Specification Test Results

Melting point 220-224°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR