Thieno[2,3-d]pyrimidin-4-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Aminothieno[2,3-d]pyrimidine, Thieno[2,3-d]pyrimidin-4-amine

044838-500MG 044838-1G
101867-818EA 318.46 USD
101867-818 101867-820
Thieno[2,3-d]pyrimidin-4-ylamine ≥95%
Thieno[2,3-d]pyrimidin-4-ylamine

Hazard: Irritant

Formula: C₆H₅N₃S
MW: 151.19 g/mol
Melting Pt: 234…236 °C
MDL Number: MFCD01316189
CAS Number: 14080-56-9

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR