2-sulfanyl-5,6,7,8-tetrahydro-4-quinazolinol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-5,6,7,8-tetrahydroquinazoline-2-thiol

044834-500MG 044834-1G 044834-5G
101867-808EA 204.62 USD
101867-808 101867-810 101867-812
2-sulfanyl-5,6,7,8-tetrahydro-4-quinazolinol ≥95%
2-sulfanyl-5,6,7,8-tetrahydro-4-quinazolinol

Hazard: Irritant

Formula: C₈H₁₀N₂OS
MW: 182.25 g/mol
Melting Pt: 337…339 °C
MDL Number: MFCD00665849
CAS Number: 16064-21-4

Order Now

Specification Test Results

Melting point 337-339°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR