4-Phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Phenyl-1H-1,2,4-triazol-5(4H)-one

044833-500MG 044833-1G 044833-5G
101867-802EA 263.08 USD
101867-802 101867-804 101867-806
4-Phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%
4-Phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one

Hazard: Irritant

Formula: C₈H₇N₃O
MW: 161.16 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00266689
CAS Number: 1008-30-6

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR