5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole ≥95%

Supplier: Matrix Scientific

044827-500MG 044827-1G 044827-5G
101867-778EA 153.85 USD
101867-778 101867-780 101867-782
5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole ≥95%
5-(Chloromethyl)-3-(4-methoxyphenyl)isoxazole

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
MW: 223.66 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD01935948
CAS Number: 325744-41-0

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR