5-(Chloromethyl)-3-phenylisoxazole ≥95%

Supplier: Matrix Scientific

044826-500MG 044826-1G 044826-5G
101867-772EA 556.92 USD
101867-772 101867-774 101867-776
5-(Chloromethyl)-3-phenylisoxazole ≥95%
5-(Chloromethyl)-3-phenylisoxazole

Hazard: Irritant

Formula: C₁₀H₈ClNO
Melting Pt: 68…70 °C
MDL Number: MFCD01935949
CAS Number: 1011-37-6

Order Now

Specification Test Results

Melting point 68-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR