2-Amino-5-chloroquinazoline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloroquinazolin-2-amine

044824-500MG 044824-1G 044824-5G
101867-760EA 115.38 USD
101867-760 101867-762 101867-764
2-Amino-5-chloroquinazoline ≥95%
2-Amino-5-chloroquinazoline

Hazard: Irritant

Formula: C₈H₆ClN₃
MW: 179.606 g/mol
Melting Pt: 249…252 °C
MDL Number: MFCD00974335
CAS Number: 190273-70-2

Order Now

Specification Test Results

Melting point 249-252°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR