2-(Methylsulfinyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

044822-500MG 044822-1G 044822-5G
101867-754EA 346.15 USD
101867-754 101867-756 101867-758
2-(Methylsulfinyl)benzenecarboxylic acid ≥95%
2-(Methylsulfinyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₈H₈O₃S
Melting Pt: 166…170 °C
MDL Number: MFCD01936003
CAS Number: 19093-34-6

Order Now

Specification Test Results

Melting point 166-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR