6-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044820-500MG 044820-1G 044820-5G
101867-742EA 556.92 USD
101867-742 101867-744 101867-746
6-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%
6-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClNOS
Melting Pt: 61.5…63.5 °C
MDL Number: MFCD00974900
CAS Number: 328547-40-6

Order Now

Specification Test Results

Melting point 61.5-63.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR