6-(4-Methylphenoxy)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044819-500MG 044819-1G 044819-5G
101867-736EA 556.92 USD
101867-736 101867-738 101867-740
6-(4-Methylphenoxy)nicotinaldehyde ≥95%
6-(4-Methylphenoxy)nicotinaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
Melting Pt: 66…68 °C
MDL Number: MFCD00974899
CAS Number: 338960-65-9

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR