(6-Chloro-3-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

044817-5G 044817-25G
101867-734EA 53.85 USD
101867-734 102551-334
(6-Chloro-3-pyridinyl)methanol ≥95%
(6-Chloro-3-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 47…49 °C
MDL Number: MFCD01075226
CAS Number: 21543-49-7

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR