5-Amino-2,4-dichlorophenol ≥95%

Supplier: Matrix Scientific

044816-500MG 044816-1G
101867-726EA 278.46 USD
101867-726 101867-728
5-Amino-2,4-dichlorophenol ≥95%
5-Amino-2,4-dichlorophenol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD00186187
CAS Number: 39489-79-7

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR