5-(Benzyloxy)-2,4-dichloroaniline ≥95%

Supplier: Matrix Scientific

044814-500MG 044814-1G
101867-716EA 556.92 USD
101867-716 101867-718
5-(Benzyloxy)-2,4-dichloroaniline ≥95%
5-(Benzyloxy)-2,4-dichloroaniline

Hazard: Irritant

Formula: C₁₃H₁₁Cl₂NO
Melting Pt: 102…105 °C
MDL Number: MFCD00974417
CAS Number: 338960-25-1

Order Now

Specification Test Results

Melting point 102-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR