Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

044813-500MG 044813-1G 044813-5G
101867-710EA 278.46 USD
101867-710 101867-712 101867-714
Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₆FNO₃
MW: 277.3 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD00664293
CAS Number: 303139-43-7

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR